Gene Transcripts
- Gene Symbol
- VENTXP1
- Entrez Gene ID
- 139538
- Gene Name
- VENT homeobox pseudogene 1
- Gene Aliases
- CT18, NA88A, VENTX2P1
- Chromosome Location
- Chr. X: 26558337 - 26561052 on Build GRCh38
- UniGene ID
- Hs.447377
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001559.2 |
|
1 |
2079 |
|
View Full Product Details