Gene Transcripts
- Gene Symbol
- FAM86HP
- Entrez Gene ID
- 729375
- Gene Name
- family with sequence similarity 86, member A pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 3: 130097782 - 130111433 on Build GRCh38
- UniGene ID
- Hs.123633
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024252.1 |
|
3 |
370 |
|
GenBank mRNA |
DA296336.1 |
- |
3 |
370 |
|
View Full Product Details