Gene Transcripts
- Gene Symbol
- ANXA2P1
- Entrez Gene ID
- 303
- Gene Name
- annexin A2 pseudogene 1
- Gene Aliases
- ANX2L1, ANX2P1, LPC2A
- Chromosome Location
- Chr. 4: 153307469 - 153308811 on Build GRCh38
- UniGene ID
- Hs.546235
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001562.2 |
|
1 |
693 |
|
View Full Product Details