Gene Transcripts
- Gene Symbol
- ANXA2P3
- Entrez Gene ID
- 305
- Gene Name
- annexin A2 pseudogene 3
- Gene Aliases
- ANX2L3, ANX2P3, LIP2, LPC2C
- Chromosome Location
- Chr. 10: 64825528 - 64826877 on Build GRCh38
- UniGene ID
- Hs.591361
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001446.2 |
|
1 |
689 |
|
View Full Product Details