Gene Transcripts
- Gene Symbol
- HSD52
- Entrez Gene ID
- 729467
- Gene Name
- uncharacterized LOC729467
- Gene Aliases
- Chromosome Location
- Chr. 1: 59131936 - 59146807 on Build GRCh38
- UniGene ID
- Hs.662291
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027120.1 |
|
3 |
731 |
|
View Full Product Details