Gene Transcripts
- Gene Symbol
- ZSCAN12P1
- Entrez Gene ID
- 221584
- Gene Name
- zinc finger and SCAN domain containing 12 pseudogene 1
- Gene Aliases
- ZNF187p1, ZNF305P1, ZNF96L1, ZNF96P1, ZSCAN12L1, dJ313I6.7
- Chromosome Location
- Chr. 6: 28090807 - 28095715 on Build GRCh38
- UniGene ID
- Hs.634723
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024063.2 |
|
2 |
3265 |
|
GenBank mRNA |
AK127188.1 |
- |
1 |
787 |
|
BC040524.1 |
- |
2 |
2921 |
|
View Full Product Details