Gene Transcripts
- Gene Symbol
- AACSP1
- Entrez Gene ID
- 729522
- Gene Name
- acetoacetyl-CoA synthetase pseudogene 1
- Gene Aliases
- AACSL
- Chromosome Location
- Chr. 5: 178764861 - 178818435 on Build GRCh38
- UniGene ID
- Hs.382790
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024035.2 |
|
11 |
2771 |
|
NR_135095.1 |
|
11 |
2774 |
|
View Full Product Details