Gene Transcripts
- Gene Symbol
- NBEAP1
- Entrez Gene ID
- 606
- Gene Name
- neurobeachin pseudogene 1
- Gene Aliases
- BCL8, BCL8A
- Chromosome Location
- Chr. 15: 20669468 - 20756151 on Build GRCh38
- UniGene ID
- Hs.657985
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027992.1 |
|
3 |
460 |
|
GenBank mRNA |
AL832227.1 |
- |
4 |
721 |
|
DA278599.1 |
- |
3 |
460 |
|
View Full Product Details