Gene Transcripts
- Gene Symbol
- HOXA11-AS
- Entrez Gene ID
- 221883
- Gene Name
- HOXA11 antisense RNA
- Gene Aliases
- HOXA-AS5, HOXA11-AS1, HOXA11AS, HOXA11S, NCRNA00076
- Chromosome Location
- Chr. 7: 27185408 - 27189293 on Build GRCh38
- UniGene ID
- Hs.587427
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002795.2 |
|
1 |
444 |
|
GenBank mRNA |
BI753022.1 |
- |
1 |
413 |
|
View Full Product Details