Gene Transcripts
- Gene Symbol
- HCG26
- Entrez Gene ID
- 352961
- Gene Name
- HLA complex group 26 (non-protein coding)
- Gene Aliases
- 3.8-1, NCRNA00191, bCX205D4.4, bPG181B23.4
- Chromosome Location
- Chr. 6: 31471229 - 31472408 on Build GRCh38
- UniGene ID
- Hs.132807
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002812.3 |
|
1 |
936 |
|
GenBank mRNA |
L29376.1 |
- |
1 |
935 |
|
View Full Product Details