Gene Transcripts
- Gene Symbol
- HSP90AB4P
- Entrez Gene ID
- 664618
- Gene Name
- heat shock protein 90 alpha family class B member 4, pseudogene
- Gene Aliases
- HSP90Bd
- Chromosome Location
- Chr. 15: 58690473 - 58693607 on Build GRCh38
- UniGene ID
- Hs.670224
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_073415.2 |
|
1 |
937 |
|
GenBank mRNA |
AY956765.1 |
- |
1 |
455 |
|
View Full Product Details