Gene Transcripts
- Gene Symbol
- GSTTP1
- Entrez Gene ID
- 25774
- Gene Name
- glutathione S-transferase theta pseudogene 1
- Gene Aliases
- GSTT4, HS322B1A
- Chromosome Location
- Chr. 22: 23998401 - 24005361 on Build GRCh38
- UniGene ID
- Hs.652490
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003081.1 |
|
4 |
632 |
|
GenBank mRNA |
AL050257.1 |
- |
4 |
632 |
|
- Gene Symbol
- LOC100652871
- Entrez Gene ID
- 100652871
- Gene Name
- glutathione S-transferase theta-4-like
- Gene Aliases
- Chromosome Location
- Chr. 22: 23935477 - 23942861 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details