Gene Transcripts

Gene Symbol
ANAPC13
Entrez Gene ID
25847
Gene Name
anaphase promoting complex subunit 13
Gene Aliases
APC13, SWM1
Chromosome Location
Chr. 3: 134477704 - 134486023 on Build GRCh38
UniGene ID
Hs.106909
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242374.1 NP_001229303.1 1 174
NM_001242375.1 NP_001229304.1 1 140
GenBank mRNA AK001285.1 - 1 171
AL050051.1 - 1 140
CN258858.1 - 1 121


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BM459488.1 - 69 Chr. - - [-]
GenBankmRNA D52765.1 - 1 105 Chr. 3 - 134478632 - 134485954 [-]
GenBankmRNA D55213.1 - 1 105 Chr. 3 - 134478681 - 134485954 [-]
GenBankmRNA HY181801.1 - 1 137 Chr. 3 - 134478653 - 134485985 [-]
GenBankmRNA AL550326.3 - 1 123 Chr. 3 - 134478584 - 134485972 [-]
GenBankmRNA CB994657.1 - 162 Chr. - - [-]
GenBankmRNA BM807200.1 - 1 133 Chr. 3 - 134478128 - 134485982 [-]

View Full Product Details