Gene Transcripts
- Gene Symbol
- SDHAP1
- Entrez Gene ID
- 255812
- Gene Name
- succinate dehydrogenase complex flavoprotein subunit A pseudogene 1
- Gene Aliases
- SDHAL1, SDHALP1
- Chromosome Location
- Chr. 3: 195959921 - 195990279 on Build GRCh38
- UniGene ID
- Hs.566872
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003264.2 |
|
16 |
2280 |
|
GenBank mRNA |
AF088032.1 |
- |
4 |
391 |
|
View Full Product Details