Gene Transcripts
- Gene Symbol
- FAM92A1P2
- Entrez Gene ID
- 403315
- Gene Name
- family with sequence similarity 92, member A3
- Gene Aliases
- FAM92A3
- Chromosome Location
- Chr. 4: 183037665 - 183040119 on Build GRCh38
- UniGene ID
- Hs.652187
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003612.2 |
|
1 |
2236 |
|
GenBank mRNA |
BC022019.1 |
- |
1 |
2199 |
|
BC059396.1 |
- |
1 |
2236 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ429004.1 |
- |
1 |
468 |
|
Chr. 4 - 183039434 - 183040093 [+] |
View Full Product Details