Gene Transcripts
- Gene Symbol
- SNORA70B
- Entrez Gene ID
- 100124537
- Gene Name
- small nucleolar RNA, H/ACA box 70B
- Gene Aliases
- Chromosome Location
- Chr. 2: 61417244 - 61417377 on Build GRCh38
- UniGene ID
- Hs.675825
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003707.2 |
|
1 |
91 |
|
View Full Product Details