Gene Transcripts

Gene Symbol
TTTY5
Entrez Gene ID
83863
Gene Name
testis-specific transcript, Y-linked 5 (non-protein coding)
Gene Aliases
LINC00126, NCRNA00126, TTY5
Chromosome Location
Chr. Y: 22296798 - 22298876 on Build GRCh38
UniGene ID
Hs.121389
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001541.1 1 451


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AF332236.3 - 1 451 Chr. Y - 22296794 - 22298876 [-]

View Full Product Details