Gene Transcripts

Gene Symbol
TTTY14
Entrez Gene ID
83869
Gene Name
testis-specific transcript, Y-linked 14 (non-protein coding)
Gene Aliases
CYorf14, NCRNA00137, NCRNA00185, PRO2834, TTY14
Chromosome Location
Chr. Y: 18872501 - 19077547 on Build GRCh38
UniGene ID
Hs.138453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001543.3 1 490
NR_125733.1 1 490
NR_125734.1 1 490
NR_125735.1 1 490
GenBank mRNA BI754238.1 - 1 240
BI824313.1 - 1 284
BX280014.1 - 1 216
DA411985.1 - 1 490
FY211103.1 - 1 153
FY211594.1 - 1 154
FY213451.1 - 1 153


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX280417.1 - 1 338 Chr. Y - 18932704 - 19077395 [-]
GenBankmRNA AF332243.3 - 1 359 Chr. Y - 18932699 - 19077416 [-]
GenBankmRNA BI823157.1 - 62 Chr. - - [-]
GenBankmRNA HY149144.1 - 1 333 Chr. Y - 18877088 - 19077389 [-]

View Full Product Details