Gene Transcripts

Gene Symbol
RACGAP1P
Entrez Gene ID
83956
Gene Name
Rac GTPase activating protein 1 pseudogene
Gene Aliases
FKSG42
Chromosome Location
Chr. 12: 45062618 - 45065411 on Build GRCh38
UniGene ID
Hs.650500
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026583.1 1 141
GenBank mRNA AF334184.1 - 1 141
BC125190.1 - 1 132
BC125191.1 - 1 132

View Full Product Details