Gene Transcripts

Gene Symbol
KHDC1
Entrez Gene ID
80759
Gene Name
KH homology domain containing 1
Gene Aliases
C6orf147, C6orf148, Em:AC019205.8, NDG1, bA257K9.4
Chromosome Location
Chr. 6: 73241314 - 73310365 on Build GRCh38
UniGene ID
Hs.433062
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251874.1 NP_001238803.1 1 574
NR_027005.2 2 147
GenBank mRNA AK090984.1 - 1 474
BM747426.1 - 2 147


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA540857.1 - 1 416 Chr. 6 - 73275535 - 73310057 [-]
GenBankmRNA BM544101.1 - 1 525 Chr. 6 - 73242071 - 73310149 [-]
GenBankmRNA DW435058.1 - 1 101 Chr. 6 - 73291976 - 73309729 [-]
GenBankmRNA BM760106.1 - 2 147 Chr. 6 - 73275088 - 73310365 [-]
GenBankmRNA BM744218.1 - 2 148 Chr. 6 - 73275462 - 73310365 [-]
GenBankmRNA BM754816.1 - 2 147 Chr. 6 - 73275049 - 73310365 [-]

View Full Product Details