Gene Transcripts
- Gene Symbol
- ID2B
- Entrez Gene ID
- 84099
- Gene Name
- inhibitor of DNA binding 2B, HLH protein (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 3: 62123423 - 62124718 on Build GRCh38
- UniGene ID
- Hs.591670
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M96843.1 |
- |
1 |
344 |
|
View Full Product Details