Gene Transcripts

Gene Symbol
ZIC4
Entrez Gene ID
84107
Gene Name
Zic family member 4
Gene Aliases
Chromosome Location
Chr. 3: 147386048 - 147406893 on Build GRCh38
UniGene ID
Hs.415766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033118.2 1 237
NR_033119.2 1 237
NR_040762.1 1 237
GenBank mRNA AK126595.1 - 1 740
AK127733.1 - 1 199
DA106797.1 - 1 169
DA121447.1 - 1 237
DA314072.1 - 1 204
DA411998.1 - 1 168
DA416382.1 - 1 53

View Full Product Details