Gene Transcripts

Gene Symbol
SNORA64
Entrez Gene ID
26784
Gene Name
small nucleolar RNA, H/ACA box 64
Gene Aliases
RNU64, U64
Chromosome Location
Chr. 16: 1962973 - 1963106 on Build GRCh38
UniGene ID
Hs.745496
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002326.1 1 109
GenBank mRNA Y11158.1 - 1 109


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BE936551.1 - 1 261 Chr. 16 - 1962942 - 1963244 [-]
GenBankmRNA DR157078.1 - 3 535 Chr. 16 - 1962855 - 1964631 [-]
GenBankmRNA CN421741.1 - 3 525 Chr. 16 - 1962775 - 1964624 [-]
GenBankmRNA CT002357.1 - 3 515 Chr. 16 - 1962969 - 1964614 [-]
GenBankmRNA CN421747.1 - 1 261 Chr. 16 - 1962629 - 1963258 [-]
GenBankmRNA BM563662.1 - 554 Chr. - - [-]
GenBankmRNA CN405534.1 - 1 109 Chr. 16 - 1962214 - 1963096 [-]
GenBankmRNA BG282897.1 - 3 535 Chr. 16 - 1962795 - 1964631 [-]
GenBankmRNA CN421735.1 - 3 504 Chr. 16 - 1962947 - 1964602 [-]
GenBankmRNA BE263765.1 - 3 517 Chr. 16 - 1962820 - 1964616 [-]
GenBankmRNA CN405671.1 - 2 323 Chr. 16 - 1962497 - 1964333 [-]
GenBankmRNA CN421739.1 - 3 514 Chr. 16 - 1962877 - 1964613 [-]
GenBankmRNA CX781018.1 - 3 482 Chr. 16 - 1962579 - 1964581 [-]
GenBankmRNA T68031.1 - 169 Chr. - - [-]
GenBankmRNA CR996341.1 - 3 537 Chr. 16 - 1962882 - 1964631 [-]
GenBankmRNA CT004387.1 - 3 522 Chr. 16 - 1962965 - 1964621 [-]
GenBankmRNA BI910102.1 - 3 531 Chr. 16 - 1962712 - 1964631 [-]

View Full Product Details