Gene Transcripts
- Gene Symbol
- RNU5A-1
- Entrez Gene ID
- 26831
- Gene Name
- RNA, U5A small nuclear 1
- Gene Aliases
- RNU5, RNU5A, RNU5C, U5A, U5B1
- Chromosome Location
- Chr. 15: 65296051 - 65296167 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002756.2 |
|
1 |
25 |
|
GenBank mRNA |
K03167.1 |
- |
1 |
25 |
|
X04215.1 |
- |
1 |
25 |
|
View Full Product Details