Gene Transcripts
- Gene Symbol
- RNU4ATAC
- Entrez Gene ID
- 100151683
- Gene Name
- RNA, U4atac small nuclear (U12-dependent splicing)
- Gene Aliases
- MOPD1, RFMN, RNU4ATAC1, TALS, U4ATAC
- Chromosome Location
- Chr. 2: 121530880 - 121531009 on Build GRCh38
- UniGene ID
- Hs.689638
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_023343.1 |
|
1 |
95 |
|
GenBank mRNA |
U62822.1 |
- |
1 |
95 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA487662.1 |
- |
|
86 |
|
Chr. 9 - 132821060 - 133019884 [+] |
GenBankmRNA |
DA039499.1 |
- |
|
95 |
|
Chr. - - [-] |
GenBankmRNA |
DA697585.1 |
- |
|
95 |
|
Chr. - - [-] |
GenBankmRNA |
DA438209.1 |
- |
|
95 |
|
Chr. - - [-] |
GenBankmRNA |
DB018378.1 |
- |
|
95 |
|
Chr. - - [-] |
View Full Product Details