Gene Transcripts
- Gene Symbol
- NXF4
- Entrez Gene ID
- 55999
- Gene Name
- nuclear RNA export factor 4 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. X: 102549965 - 102571693 on Build GRCh38
- UniGene ID
- Hs.454283
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002216.1 |
|
4 |
484 |
|
GenBank mRNA |
AK124700.1 |
- |
4 |
484 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA187251.1 |
- |
4 |
484 |
|
Chr. X - 102549965 - 102562686 [+] |
View Full Product Details