Gene Transcripts
- Gene Symbol
- SARNP
- Entrez Gene ID
- 84324
- Gene Name
- SAP domain containing ribonucleoprotein
- Gene Aliases
- CIP29, HCC1, HSPC316, THO1
- Chromosome Location
- Chr. 12: 55752463 - 55817756 on Build GRCh38
- UniGene ID
- Hs.505676
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026722.1 |
|
11 |
1068 |
|
NR_026723.1 |
|
12 |
1115 |
|
GenBank mRNA |
AK290508.1 |
- |
12 |
1083 |
|
View Full Product Details