Gene Transcripts
- Gene Symbol
- KC6
- Entrez Gene ID
- 641516
- Gene Name
- keratoconus gene 6
- Gene Aliases
- Chromosome Location
- Chr. 18: 41480271 - 41520597 on Build GRCh38
- UniGene ID
- Hs.116612
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002838.1 |
|
6 |
1139 |
|
GenBank mRNA |
AY762618.1 |
- |
6 |
1139 |
|
View Full Product Details