Gene Transcripts
- Gene Symbol
- LINC01547
- Entrez Gene ID
- 84536
- Gene Name
- long intergenic non-protein coding RNA 1547
- Gene Aliases
- C21orf67, C21orf69, PRED54
- Chromosome Location
- Chr. 21: 44933284 - 44939913 on Build GRCh38
- UniGene ID
- Hs.534504
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
FY213399.1 |
- |
2 |
441 |
|
Chr. 21 - 44934387 - 44939873 [-] |
GenBankmRNA |
FY210788.1 |
- |
2 |
264 |
|
Chr. 21 - 44934315 - 44939873 [-] |
GenBankmRNA |
FY211216.1 |
- |
|
441 |
|
Chr. - - [-] |
GenBankmRNA |
FY213940.1 |
- |
|
264 |
|
Chr. - - [-] |
GenBankmRNA |
FY211596.1 |
- |
|
441 |
|
Chr. - - [-] |
GenBankmRNA |
FY211768.1 |
- |
2 |
264 |
|
Chr. 21 - 44934317 - 44939873 [-] |
View Full Product Details