Gene Transcripts
- Gene Symbol
- LINC00475
- Entrez Gene ID
- 158314
- Gene Name
- long intergenic non-protein coding RNA 475
- Gene Aliases
- C9orf44
- Chromosome Location
- Chr. 9: 92141467 - 92159608 on Build GRCh38
- UniGene ID
- Hs.149940
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027341.1 |
|
2 |
274 |
|
GenBank mRNA |
AK023662.1 |
- |
2 |
274 |
|
BC128384.1 |
- |
2 |
159 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AU139106.2 |
- |
2 |
274 |
|
Chr. 9 - 92141467 - 92158206 [+] |
GenBankmRNA |
BI829433.1 |
- |
2 |
126 |
|
Chr. 9 - 92152594 - 92158479 [+] |
View Full Product Details