Gene Transcripts
- Gene Symbol
- AFAP1-AS1
- Entrez Gene ID
- 84740
- Gene Name
- AFAP1 antisense RNA 1
- Gene Aliases
- AFAP1-AS, AFAP1AS
- Chromosome Location
- Chr. 4: 7754090 - 7778928 on Build GRCh38
- UniGene ID
- Hs.663029
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026892.1 |
|
2 |
5200 |
|
GenBank mRNA |
BC004397.1 |
- |
1 |
75 |
|
View Full Product Details