Gene Transcripts
- Gene Symbol
- GUSBP10
- Entrez Gene ID
- 642006
- Gene Name
- glucuronidase, beta pseudogene 10
- Gene Aliases
- Chromosome Location
- Chr. 7: 57165670 - 57180156 on Build GRCh38
- UniGene ID
- Hs.562648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_030766.1 |
|
4 |
955 |
|
View Full Product Details