Gene Transcripts
- Gene Symbol
- HBBP1
- Entrez Gene ID
- 3044
- Gene Name
- hemoglobin subunit beta pseudogene 1
- Gene Aliases
- HBH1, HBHP
- Chromosome Location
- Chr. 11: 5241954 - 5243592 on Build GRCh38
- UniGene ID
- Hs.20205
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001589.1 |
|
1 |
24 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
U01317.3 |
- |
2 |
45681 |
|
Chr. 11 - 5215910 - 5289430 [-] |
View Full Product Details