Gene Transcripts
- Gene Symbol
- SNHG7
- Entrez Gene ID
- 84973
- Gene Name
- small nucleolar RNA host gene 7
- Gene Aliases
- NCRNA00061
- Chromosome Location
- Chr. 9: 136724594 - 136728184 on Build GRCh38
- UniGene ID
- Hs.495480
- Species
- Homo sapiens
- Gene Symbol
- SNORA17B
- Entrez Gene ID
- 677824
- Gene Name
- small nucleolar RNA, H/ACA box 17B
- Gene Aliases
- ACA43, SNORA43
- Chromosome Location
- Chr. 9: 136726104 - 136726237 on Build GRCh38
- UniGene ID
- Hs.495480
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054908.1 |
- |
2 |
323 |
|
View Full Product Details