Gene Transcripts

Gene Symbol
ENSG00000186594
Entrez Gene ID
ENSG00000186594
Gene Name
Gene Aliases
Chromosome Location
Chr. : - on Build
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID

Gene Symbol
MIR22HG
Entrez Gene ID
84981
Gene Name
MIR22 host gene
Gene Aliases
C17orf91
Chromosome Location
Chr. 17: 1711504 - 1716272 on Build GRCh38
UniGene ID
Hs.597755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028502.1 2 2072
NR_028503.1 3 1265
NR_028504.1 4 852
NR_028505.1 3 769
GenBank mRNA AF070569.1 - 1 1641
BX640650.1 - 4 790
BX648321.1 - 2 2009

View Full Product Details