Gene Transcripts
- Gene Symbol
- SPATA31D5P
- Entrez Gene ID
- 347127
- Gene Name
- SPATA31 subfamily D member 5, pseudogene
- Gene Aliases
- FAM75D5, FAM75D5P
- Chromosome Location
- Chr. 9: 81913437 - 81919927 on Build GRCh38
- UniGene ID
- Hs.215235
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026851.1 |
|
4 |
1390 |
|
GenBank mRNA |
AK125938.1 |
- |
4 |
1274 |
|
AK128229.1 |
- |
4 |
1887 |
|
View Full Product Details