Gene Transcripts

Gene Symbol
TDRG1
Entrez Gene ID
732253
Gene Name
testis development related 1 (non-protein coding)
Gene Aliases
LINC00532
Chromosome Location
Chr. 6: 40378424 - 40379892 on Build GRCh38
UniGene ID
Hs.180197
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024015.1 2 778
GenBank mRNA BC033995.2 - 2 865
BC042123.1 - 2 865
DQ168992.1 - 2 778


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA293846.1 - 2 491 Chr. 6 - 40378711 - 40379597 [+]
GenBankmRNA BG183126.1 - 463 Chr. - - [-]
GENCODE ENST00000448559.5 - 2 441 Chr. 6 - 40340465 - 40379885 [+]
GENCODE ENST00000451810.2 - 3 617 Chr. 6 - 40334954 - 40379885 [+]
GENCODE ENST00000373170.2 - 2 865 Chr. 6 - 40378337 - 40379899 [+]

View Full Product Details