Gene Transcripts
- Gene Symbol
- SCARNA2
- Entrez Gene ID
- 677766
- Gene Name
- small Cajal body-specific RNA 2
- Gene Aliases
- HBII-382, mgU2-25/61
- Chromosome Location
- Chr. 1: 109100193 - 109100612 on Build GRCh38
- UniGene ID
- Hs.550815
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003023.1 |
|
1 |
357 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA439976.1 |
- |
1 |
357 |
|
Chr. 1 - 109100195 - 109100774 [+] |
GenBankmRNA |
BU564533.1 |
- |
|
360 |
|
Chr. - - [-] |
View Full Product Details