Gene Transcripts
- Gene Symbol
- SCARNA22
- Entrez Gene ID
- 677770
- Gene Name
- small Cajal body-specific RNA 22
- Gene Aliases
- ACA11
- Chromosome Location
- Chr. 4: 1974636 - 1974760 on Build GRCh38
- UniGene ID
- Hs.676951
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003004.1 |
|
1 |
115 |
|
GenBank mRNA |
AJ609470.1 |
- |
1 |
115 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV744827.1 |
- |
|
161 |
|
Chr. - - [-] |
View Full Product Details