Gene Transcripts
- Gene Symbol
- JHDM1D-AS1
- Entrez Gene ID
- 100134229
- Gene Name
- JHDM1D antisense RNA 1 (head to head)
- Gene Aliases
- Chromosome Location
- Chr. 7: 140177261 - 140179640 on Build GRCh38
- UniGene ID
- Hs.634333
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024451.1 |
|
1 |
1759 |
|
GenBank mRNA |
AK026372.1 |
- |
1 |
1665 |
|
View Full Product Details