Gene Transcripts
- Gene Symbol
- RBMY2EP
- Entrez Gene ID
- 159125
- Gene Name
- RNA binding motif protein, Y-linked, family 2, member E pseudogene
- Gene Aliases
- Chromosome Location
- Chr. Y: 21395148 - 21401562 on Build GRCh38
- UniGene ID
- Hs.567745
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001574.2 |
|
3, 4 |
301 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
U94388.3 |
- |
3, 4 |
301 |
|
Chr. Y - 21395148 - 21401562 [-] |
View Full Product Details