Gene Transcripts

Gene Symbol
PCAT4
Entrez Gene ID
118425
Gene Name
prostate cancer associated transcript 4 (non-protein coding)
Gene Aliases
GDEP, PCA4, PCAN1
Chromosome Location
Chr. 4: 79827471 - 79863247 on Build GRCh38
UniGene ID
Hs.104215
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026555.1 2 283
GenBank mRNA AF388200.1 - 2 278
AF462307.1 - 2 234
AK056825.1 - 2 283
BC105591.1 - 2 381
BC125139.1 - 2 280

View Full Product Details