Gene Transcripts
- Gene Symbol
- ARGFXP2
- Entrez Gene ID
- 503640
- Gene Name
- arginine-fifty homeobox pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 17: 32150357 - 32151571 on Build GRCh38
- UniGene ID
- Hs.631752
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002222.1 |
|
1 |
307 |
|
View Full Product Details