Gene Transcripts

Gene Symbol
SNORA32
Entrez Gene ID
692063
Gene Name
small nucleolar RNA, H/ACA box 32
Gene Aliases
ACA32
Chromosome Location
Chr. 11: 93730979 - 93731099 on Build GRCh38
UniGene ID
Hs.738740
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003032.1 1 79

Gene Symbol
TAF1D
Entrez Gene ID
79101
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit D
Gene Aliases
JOSD3, RAFI41, TAF(I)41, TAFI41
Chromosome Location
Chr. 11: 93735929 - 93741537 on Build GRCh38
UniGene ID
Hs.355750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK027016.1 - 12 1475
AK057183.1 - 1 1820
AK128061.1 - 3 3848

View Full Product Details