Gene Transcripts
- Gene Symbol
- FUNDC2P2
- Entrez Gene ID
- 388965
- Gene Name
- FUN14 domain containing 2 pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 2: 84290682 - 84292200 on Build GRCh38
- UniGene ID
- Hs.664098
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003663.1 |
|
1 |
127 |
|
GenBank mRNA |
BC067852.1 |
- |
1 |
126 |
|
View Full Product Details