Gene Transcripts
- Gene Symbol
- EGOT
- Entrez Gene ID
- 100126791
- Gene Name
- eosinophil granule ontogeny transcript (non-protein coding)
- Gene Aliases
- EGO, NCRNA00190
- Chromosome Location
- Chr. 3: 4749192 - 4751590 on Build GRCh38
- UniGene ID
- Hs.663059
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_004428.1 |
|
2 |
1013 |
|
GenBank mRNA |
BC039547.1 |
- |
2 |
1013 |
|
View Full Product Details