Gene Transcripts

Gene Symbol
CYMP
Entrez Gene ID
643160
Gene Name
chymosin pseudogene
Gene Aliases
Chromosome Location
Chr. 1: 110480766 - 110491269 on Build GRCh38
UniGene ID
Hs.532720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003599.2 6 730
GenBank mRNA AK310315.1 - 8 1126


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX096065.1 - 1 75 Chr. 1 - 110488529 - 110491270 [+]
GenBankmRNA CN284362.1 - 4 504 Chr. 1 - 110480700 - 110488783 [+]
GenBankmRNA AA411566.1 - 1 77 Chr. 1 - 110488527 - 110491265 [+]

View Full Product Details