Gene Transcripts
- Gene Symbol
- TREML3P
- Entrez Gene ID
- 340206
- Gene Name
- triggering receptor expressed on myeloid cells like 3, pseudogene
- Gene Aliases
- TLT3, TREML3
- Chromosome Location
- Chr. 6: 41208554 - 41217947 on Build GRCh38
- UniGene ID
- Hs.93698
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027256.1 |
|
5 |
951 |
|
GenBank mRNA |
AF534825.1 |
- |
5 |
951 |
|
View Full Product Details