Gene Transcripts

Gene Symbol
SPDYE8P
Entrez Gene ID
728524
Gene Name
speedy/RINGO cell cycle regulator family member E8, pseudogene
Gene Aliases
Chromosome Location
Chr. 7: 73019731 - 73029762 on Build GRCh38
UniGene ID
Hs.571275
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003664.2 6 1247
GenBank mRNA BC045636.1 - 6 1250
BC068610.1 - 6 1250
BC110795.1 - 2 514

Gene Symbol
SPDYE11
Entrez Gene ID
100996746
Gene Name
speedy/RINGO cell cycle regulator family member E11
Gene Aliases
Chromosome Location
Chr. 7: 73049256 - 73056645 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006716210.3 XP_006716273.1 5 1206
XM_017012933.1 XP_016868422.1 5 1210

Gene Symbol
SPDYE17
Entrez Gene ID
102723849
Gene Name
speedy/RINGO cell cycle regulator family member E17
Gene Aliases
Chromosome Location
Chr. 7: 77023682 - 77031104 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006716212.3 XP_006716275.1 5 1210
XM_017012888.1 XP_016868377.1 5 1210

View Full Product Details