Gene Transcripts
- Gene Symbol
- PWRN1
- Entrez Gene ID
- 791114
- Gene Name
- Prader-Willi region non-protein coding RNA 1
- Gene Aliases
- NCRNA00198
- Chromosome Location
- Chr. 15: 24558157 - 24587780 on Build GRCh38
- UniGene ID
- Hs.720900
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026646.1 |
|
7, 8 |
1121 |
|
GenBank mRNA |
BC034815.1 |
- |
7, 8 |
1121 |
|
View Full Product Details